Anti-USP25 Rabbit Polyclonal Antibody

Supplier: ProSci
7765_S
10748-904EA 185.24 USD
10748-904
Anti-USP25 Rabbit Polyclonal Antibody
Antibodies
USP25 (ubiquitin specific peptidase 25), also known as USP21, belongs to the peptidase C19 family and is a highly conserved 76-amino acid protein involved in regulation of intracellular protein breakdown, cell cycle regulation, and stress response. It contains one UBA-like domain and two UIM (ubiquitin-interacting motif) repeats. Due to alternative splicing events, USP25 is expressed as two short, ubiquitously expressed isoforms and one long, muscle-specific isoform. The long isoform of USP25 (USP25m) is upregulated in myogenesis and is implicated in regulation of muscular differentiation and function. USP25 is a deubiquitinating enzyme (DUB) that negatively regulates IL-17-triggered signaling.

Type: Primary
Antigen: USP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR