Ward's® Datahub, General Science With Charging Pins
470217-544KT 625 USD
470217-544
Ward's® Datahub, General Science With Charging Pins
Put the power of thirteen sensors in the palm of your hand.
  • Free and easy-to-use software
  • Lesson plans and sample data tables included
  • Built in charging pins for use with the Datahub storage cart
  • Sample memory: 128K
  • Size: 132 Ø×45 H mm
  • Stopwatch: 0.1 s to 99:59.9 minutes
  • With keyboard, remote data collection, automatic sensor testing and calibration
Charging, storage, and data collection has never been easier. The new General Science Datahub now includes charging and storage docking capabilities. With Ward’s Datahub Mobile Charging Cart, users can now easily charge and store this redesigned probeware all in one place, to hold hands-on STEM-based science lessons anywhere in the classroom.
Connect real scientific processes and core concepts with the real world using this versatile and comprehensive unit, which covers a broad range of major science disciplines. With this complete system, students can measure and analyze data models for a variety of standards-based topics including traveling speed with GPS, Newton’s Laws, sound waves, electrical currents, pH titration, endothermic and exothermic reactions, Boyle’s Law, specific heat, and microclimate. Unit has 24k/sec sampling rate, 100000/s maximum sampling speed and it communicates through wireless bluetooth V2.0+EDR and USB V2.0., long-lasting battery life. Includes Ward's DataHub Software, AC/DC adapter, Quick Start Guide, and USB cable. Accessories included with this unit include banana cables, external temperature probe, air pressure extension tube, pH electrode, and plastic rod, stand, and screw. Compatible with PC/Mac and iPad devices.
Thirteen built-in sensors include Air pressure, ambient temperature, current, distance, external temperature, GPS, light, microphone, pH, relative humidity, sound level, , voltage, and universal sensor input.
Certifications: CSA approved.