Peracetic acid 35%
Supplier: Pfaltz & Bauer
|
Danger
|
Synonyms:
Peroxyacetic acid
P05020-100ML
101184-388EA
613.05
USD
101184-388
Peracetic acid 35%
Peracetic acid
Formula:
C₂H₄O₃ MW: 76.051 g/mol Boiling Pt: 107 °C Density: 1,13 Flash Pt: 46 °C |
MDL Number:
MFCD00002128 CAS Number: 79-21-0 UN: 3109 ADR: 5.2,8,3, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...