MKF 2100 Cool Unit, Built In, Medite
02-2100-00 02-2100-00
10768-894EA 4613.77 USD
10768-894 CA10768-248
MKF 2100 Cool Unit, Built In, Medite
The MKF 20 is a unique cfc-free cool unit for counter top built in.
The space-saving model offers optimal conditions when sectioning at the microtome. The digital temperature control with digital display at the lower part of the unit can be turned 90°, enabling, installation in longitudinal or lateral direction.