Bolton Broth Selective Supplement, MilliporeSigma
1.00079.0010
EM1.00079.0010EA 440.42 USD
EM1.00079.0010
Bolton Broth Selective Supplement, MilliporeSigma
  • 10 to 100 inoculum on reference medium Campylobacter coli ATCC 43478 (WDCM 00004)
  • 10 to 100 inoculum on reference medium Campylobacter jejuni ATCC 33291 (WDCM 00005)
  • Less than or equal to 1000 inoculum on reference medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Less than or equal to 1000 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
  • Less than or equal to 1000 inoculum on reference medium Proteus mirabilis ATCC 29906 (WDCM 00023)
  • Less than 10 CFU growth on mCCDA Campylobacter coli ATCC 43478 (WDCM 00004)
  • Less than 10 CFU growth on mCCDA Campylobacter jejuni ATCC 33291 (WDCM 00005)
  • Less than 10 CFU growth on mCCDA Campylobacter jejuni ATCC 29428 (WDCM 00156)
  • No growth on Escherichia coli ATCC 25922 (WDCM 00013)
  • No growth on Escherichia coli ATCC 8739 (WDCM 00012)
  • No growth on Proteus mirabilis ATCC 29906 (WDCM 00023)
  • Greyish colony color on Campylobacter coli ATCC 43478 (WDCM 00004)
  • Greyish colony color on Campylobacter jejuni ATCC 33291 (WDCM 00005)
  • Greyish colony color on Campylobacter jejuni ATCC 29428 (WDCM 00156)
In North America 1.00079.0010 replaces and is identical to 1.00079.0017