Anti-CSRNP2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89020
89297-074EA 607.87 USD
89297-074
Anti-CSRNP2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to TAIP-12 / FAM130A1

Approx 60kDa band observed in Human Heart and Mouse Heart lysates (calculated MW of 58.5kDa according to NP_700456.1). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: CSRNP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR