Anti-LIPG Mouse Monoclonal Antibody [clone: 7C2]

Supplier: GeneTex
GTX84193
89273-180EA 705.34 USD
89273-180
Anti-LIPG Mouse Monoclonal Antibody [clone: 7C2]
Antibodies
Mouse monoclonal antibody [7C2] to LIPG

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LIPG
Clonality: Monoclonal
Clone: 7C2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR