Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1F6]

GTX84104
89273-004EA 511.34 USD
89273-004
Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1F6]
Mouse monoclonal antibody [1F6] to MOBKL1A

Recommended Starting Dilutions: WB 1:500 IHC 1:50,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MOBKL1A
Clonality: Monoclonal
Clone: 1F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR