Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 3G9]

Supplier: GeneTex
GTX84103
89273-002EA 705.1 USD
89273-002
Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 3G9]
Antibodies
Mouse monoclonal antibody [3G9] to MOBKL1A

Recommended Starting Dilutions: WB 1:500 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MOBKL1A
Clonality: Monoclonal
Clone: 3G9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR