Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 6C6]

GTX84102
89273-000EA 450.84 USD
89273-000
Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 6C6]
Mouse monoclonal antibody [6C6] to MOBKL2B

Recommended Starting Dilutions:WB 1:2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MOBKL2B
Clonality: Monoclonal
Clone: 6C6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR