Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 7G2]

Supplier: GeneTex
GTX84099
89272-994EA 450.84 USD
89272-994
Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 7G2]
Mouse monoclonal antibody [7G2] to MOBKL2B

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MOBKL2B
Clonality: Monoclonal
Clone: 7G2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR