Anti-MRI1 Mouse Monoclonal Antibody [clone: 9H7]

Supplier: GeneTex
GTX84085
89272-966EA 705.34 USD
89272-966
Anti-MRI1 Mouse Monoclonal Antibody [clone: 9H7]
Antibodies
Mouse monoclonal antibody [9H7] to MRI1

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:1,000Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MRI1
Clonality: Monoclonal
Clone: 9H7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR