Anti-MSMB Mouse Monoclonal Antibody [clone: 6C7]

Supplier: GeneTex
GTX84083
89272-962EA 705.72 USD
89272-962
Anti-MSMB Mouse Monoclonal Antibody [clone: 6C7]
Antibodies
Mouse monoclonal antibody [6C7] to MSMB

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MSMB
Clonality: Monoclonal
Clone: 6C7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR