Anti-MTMR14 Mouse Monoclonal Antibody [clone: 7B2]

GTX84082
89272-960EA 450.84 USD
89272-960
Anti-MTMR14 Mouse Monoclonal Antibody [clone: 7B2]
Mouse monoclonal antibody [7B2] to MTMR14

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MTMR14
Clonality: Monoclonal
Clone: 7B2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR