Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MTMR14 Mouse Monoclonal Antibody [clone: 7F2]

Supplier: Genetex
GTX84081
89272-958
Anti-MTMR14 Mouse Monoclonal Antibody [clone: 7F2]
Mouse monoclonal antibody [7F2] to MTMR14

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MTMR14
Clonality: Monoclonal
Clone: 7F2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty