Anti-MTMR14 Mouse Monoclonal Antibody [clone: 6B6]

Supplier: GeneTex
GTX84079
89272-954EA 705.72 USD
89272-954
Anti-MTMR14 Mouse Monoclonal Antibody [clone: 6B6]
Antibodies
Mouse monoclonal antibody [6B6] to MTMR14

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MTMR14
Clonality: Monoclonal
Clone: 6B6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR