Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1C5]

Supplier: GeneTex
GTX84077
89272-950EA 450.84 USD
89272-950
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1C5]
Mouse monoclonal antibody [1C5] to MTRF1L

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 1C5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR