Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1000]

GTX84068
89272-932EA 511.34 USD
89272-932
Anti-MTRF1L Mouse Monoclonal Antibody [clone: 1000]
Mouse monoclonal antibody [1E3] to MTRF1L

Recommended Starting Dilutions:WB 1:1000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MTRF1L
Clonality: Monoclonal
Clone: 1000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR