Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 3F2]

Supplier: GeneTex
GTX84065
89272-926EA 705.1 USD
89272-926
Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 3F2]
Antibodies
Mouse monoclonal antibody [3F2] to c-Myc

Recommended Starting Dilutions:Western blot: 1:1000 ICC/IF (1:100)Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: 3F2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR