Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 5G3]

Supplier: GeneTex
GTX84064
89272-924EA 704.88 USD
89272-924
Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 5G3]
Antibodies
Mouse monoclonal antibody [5G3] to c-Myc

Recommended Starting Dilutions:Western blot: 1:100 ICC/IF (1:100)Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: 5G3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR