Anti-NAT8 Mouse Monoclonal Antibody [clone: 5A8]

Supplier: GeneTex
GTX84063
89272-922EA 705.34 USD
89272-922
Anti-NAT8 Mouse Monoclonal Antibody [clone: 5A8]
Antibodies
Mouse monoclonal antibody [5A8] to NAT8

Recommended Starting Dilutions:WB 1:500 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NAT8
Clonality: Monoclonal
Clone: 5A8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR