Anti-NEK11 Mouse Monoclonal Antibody [clone: 5E9]

Supplier: GeneTex
GTX84061
89272-918EA 705.72 USD
89272-918
Anti-NEK11 Mouse Monoclonal Antibody [clone: 5E9]
Antibodies
Mouse monoclonal antibody [5E9] to NEK11

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEK11
Clonality: Monoclonal
Clone: 5E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR