Anti-NEK6 Mouse Monoclonal Antibody [clone: 5B9]

Supplier: GeneTex
GTX84057
89272-910EA 450.84 USD
89272-910
Anti-NEK6 Mouse Monoclonal Antibody [clone: 5B9]
Mouse monoclonal antibody [5B9] to NEK6

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEK6
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR