Anti-NEK6 Mouse Monoclonal Antibody [clone: 8G5]

Supplier: GeneTex
GTX84056
89272-908EA 705.34 USD
89272-908
Anti-NEK6 Mouse Monoclonal Antibody [clone: 8G5]
Antibodies
Mouse monoclonal antibody [8G5] to NEK6

Recommended Starting Dilutions:WB 1:1000 IHC 1:50 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEK6
Clonality: Monoclonal
Clone: 8G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR