Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 3F9]

Supplier: GeneTex
GTX84053
89272-902EA 450.84 USD
89272-902
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 3F9]
Mouse monoclonal antibody [3F9] to NEUROG1

Recommended Starting Dilutions:WB 1:1000 IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR