Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 19A11]

Supplier: GeneTex
GTX84050
89272-896EA 450.84 USD
89272-896
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 19A11]
Mouse monoclonal antibody [19A11] to NEUROG1

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 19A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR