Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 3B5 ]

Supplier: GeneTex
GTX84046
89272-888EA 511.34 USD
89272-888
Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 3B5 ]
Mouse monoclonal antibody [3B5 ] to EUROG3

Recommended Starting Dilutions:Western blot: 1:500Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NEUROG3
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR