Anti-NHEJ1 Mouse Monoclonal Antibody [clone: 3E5]

Supplier: GeneTex
GTX84044
89272-884EA 705.1 USD
89272-884
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: 3E5]
Antibodies
Mouse Monoclonal antibody [3E5] to XLF

Recommended Starting Dilutions: WB 1:2000,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NHEJ1
Clonality: Monoclonal
Clone: 3E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR