Anti-NIT1 Mouse Monoclonal Antibody [clone: 3A11]

Supplier: GeneTex
GTX84041
89272-878EA 705.72 USD
89272-878
Anti-NIT1 Mouse Monoclonal Antibody [clone: 3A11]
Antibodies
Mouse monoclonal antibody [3A11] to NIT1

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NIT1
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR