Anti-NIT2 Mouse Monoclonal Antibody [clone: 6H7]

Supplier: GeneTex
GTX84040
89272-876EA 705.1 USD
89272-876
Anti-NIT2 Mouse Monoclonal Antibody [clone: 6H7]
Antibodies
Mouse monoclonal antibody [6H7] to NIT2

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NIT2
Clonality: Monoclonal
Clone: 6H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR