Anti-NIT2 Mouse Monoclonal Antibody [clone: 5A3]

Supplier: GeneTex
GTX84037
89272-870EA 450.84 USD
89272-870
Anti-NIT2 Mouse Monoclonal Antibody [clone: 5A3]
Mouse monoclonal antibody [5A3] to NIT2

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NIT2
Clonality: Monoclonal
Clone: 5A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR