Anti-NIT2 Mouse Monoclonal Antibody [clone: 1A6]

Supplier: GeneTex
GTX84035
89272-866EA 450.84 USD
89272-866
Anti-NIT2 Mouse Monoclonal Antibody [clone: 1A6]
Mouse monoclonal antibody [1A6] to NIT2

Recommended Starting Dilutions: WB 1:1000 IHC 1:50,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NIT2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR