Anti-NME4 Mouse Monoclonal Antibody [clone: 1000]

GTX84025
89272-846EA 450.84 USD
89272-846
Anti-NME4 Mouse Monoclonal Antibody [clone: 1000]
Mouse monoclonal antibody [1E3] to NME4

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NME4
Clonality: Monoclonal
Clone: 1000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR