Anti-NPR3 Mouse Monoclonal Antibody [clone: 11B6]

GTX84017
89272-832EA 450.84 USD
89272-832
Anti-NPR3 Mouse Monoclonal Antibody [clone: 11B6]
Mouse monoclonal antibody [11B6] to NPR3

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NPR3
Clonality: Monoclonal
Clone: 11B6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR