Anti-NPR3 Mouse Monoclonal Antibody [clone: 4C3]

Supplier: GeneTex
GTX84010
89272-818EA 735.91 USD
89272-818
Anti-NPR3 Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Mouse monoclonal antibody [4C3] to NPR3

Recommended Starting Dilutions: WB 1:2000, IHC 1:50, ICC/IF 1:100, FACS: 1:100.

Type: Primary
Antigen: NPR3
Clonality: Monoclonal
Clone: 4C3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR