Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2A5]

Supplier: GeneTex
GTX84002
89272-802EA 450.84 USD
89272-802
Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2A5]
Mouse monoclonal antibody [2A5] to NRBP1

Recommended Starting Dilutions:WB 1:1000 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NRBP1
Clonality: Monoclonal
Clone: 2A5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR