Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 4A2]

GTX83998
89272-794EA 450.84 USD
89272-794
Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 4A2]
Mouse monoclonal antibody [4A2] to NSMCE2

Recommended Starting Dilutions: WB 1:500 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NSMCE2
Clonality: Monoclonal
Clone: 4A2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR