Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 3C10]

GTX83997
89272-792EA 450.84 USD
89272-792
Anti-NSMCE2 Mouse Monoclonal Antibody [clone: 3C10]
Mouse monoclonal antibody [3C10] to NSMCE2

Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NSMCE2
Clonality: Monoclonal
Clone: 3C10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR