Anti-NT5DC1 Mouse Monoclonal Antibody [clone: 1G10]

GTX83990
89272-778EA 450.84 USD
89272-778
Anti-NT5DC1 Mouse Monoclonal Antibody [clone: 1G10]
Mouse monoclonal antibody [1G10] to NT5DC1

Recommended Starting Dilutions: WB 1:2000 IHC 1:150 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NT5DC1
Clonality: Monoclonal
Clone: 1G10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR