Anti-NT3 Mouse Monoclonal Antibody [clone: 5A2]

GTX83986
89272-770EA 511.34 USD
89272-770
Anti-NT3 Mouse Monoclonal Antibody [clone: 5A2]
Mouse monoclonal antibody [5A2] to NT3

Recommended Starting Dilutions:Western blot: 1:5000 IHC 1:50 ICC/IF (1:100)Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NT3
Clonality: Monoclonal
Clone: 5A2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR