Anti-NT3 Mouse Monoclonal Antibody [clone: 3D4]

Supplier: GeneTex
GTX83984
89272-766EA 693.22 USD
89272-766
Anti-NT3 Mouse Monoclonal Antibody [clone: 3D4]
Antibodies
Mouse monoclonal antibody [3D4] to NT3

Recommended Starting Dilutions:Western blot: 1:10000 IHC-P : 1:50,<br/>Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NT3
Clonality: Monoclonal
Clone: 3D4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR