Anti-NT3 Mouse Monoclonal Antibody [clone: 5C7]

Supplier: GeneTex
GTX83983
89272-764EA 705.1 USD
89272-764
Anti-NT3 Mouse Monoclonal Antibody [clone: 5C7]
Antibodies
Mouse monoclonal antibody [5C7] to NT3

Recommended Starting Dilutions:Western blot: 1:5000 IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NT3
Clonality: Monoclonal
Clone: 5C7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR