Anti-NTRK3 Mouse Monoclonal Antibody [clone: 24B3]

GTX83979
89272-756EA 450.84 USD
89272-756
Anti-NTRK3 Mouse Monoclonal Antibody [clone: 24B3]
Mouse monoclonal antibody [24B3] to NTRK3

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NTRK3
Clonality: Monoclonal
Clone: 24B3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR