Anti-NXNL2 Mouse Monoclonal Antibody [clone: 1C3]

Supplier: GeneTex
GTX83970
89272-738EA 705.1 USD
89272-738
Anti-NXNL2 Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
Mouse monoclonal antibody [1C3] to NXNL2

Recommended Starting Dilutions: WB 1:1000~2000 IHC 1:50 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NXNL2
Clonality: Monoclonal
Clone: 1C3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR