Anti-ODC1 Mouse Monoclonal Antibody [clone: 1G6]

Supplier: GeneTex
GTX83968
89272-734EA 735.33 USD
89272-734
Anti-ODC1 Mouse Monoclonal Antibody [clone: 1G6]
Antibodies
Mouse monoclonal antibody [1G6] to ODC1

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: 1G6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR