Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 2F6]

GTX83964
89272-726EA 450.84 USD
89272-726
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 2F6]
Mouse monoclonal antibody [2F6] to OSBPL11

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OSBPL11
Clonality: Monoclonal
Clone: 2F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR