Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 6H9]

Supplier: GeneTex
GTX83963
89272-724EA 705.1 USD
89272-724
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: 6H9]
Antibodies
Mouse monoclonal antibody [6H9] to OSBPL11

Recommended Starting Dilutions: WB 1:2000 IHC 1:150 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OSBPL11
Clonality: Monoclonal
Clone: 6H9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR