Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11B3]

Supplier: GeneTex
GTX83953
89272-704EA 706.05 USD
89272-704
Anti-OTUB2 Mouse Monoclonal Antibody [clone: 11B3]
Antibodies
Mouse monoclonal antibody [11B3] to OTUB2

Recommended Starting Dilutions: WB 1:500~2000 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OTUB2
Clonality: Monoclonal
Clone: 11B3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR