Anti-OXSR1 Mouse Monoclonal Antibody [clone: 1F3]

Supplier: GeneTex
GTX83951
89272-700EA 705.72 USD
89272-700
Anti-OXSR1 Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Mouse monoclonal antibody [1F3] to OXSR1

Recommended Starting Dilutions:WB 1:2000; IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OXSR1
Clonality: Monoclonal
Clone: 1F3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR