Anti-PANK2 Mouse Monoclonal Antibody [clone: 4B3]

GTX83945
89272-688EA 450.84 USD
89272-688
Anti-PANK2 Mouse Monoclonal Antibody [clone: 4B3]
Mouse monoclonal antibody [4B3] to PANK2

Recommended Starting Dilutions:WB 1:2000 IHC 1:150 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PANK2
Clonality: Monoclonal
Clone: 4B3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR